Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) potassium sulfate dodecahydrate, 98%

Catalog No. p-7081722
Click to view available options
Quantity:
100 g
500 g
2500 g

Catalog No. AAA1181622

Supplier: Thermo Scientific Chemicals A1181622

Only null left
Add to Cart

CAS: 7788-99-0 | CrH24KO20S2 | 499.386 g/mol

Chromium(III) potassium sulfate is used for tanning of leather and as a photographic film hardener. It is used in manufacturing of chromium-containing catalyst.It is used in mordantin in printing and dyeing industry and it could also be used as analytical reagent. In addition, it is also used as food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromium(III) potassium sulfate is used for tanning of leather and as a photographic film hardener. It is used in manufacturing of chromium-containing catalyst.It is used in mordantin in printing and dyeing industry and it could also be used as analytical reagent. In addition, it is also used as food additive.

Solubility
Soluble in water. 24 g/100 mL (15°C)

Notes
Incompatible with magnesium, aluminum. Store away from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7788-99-0
Molecular Formula CrH24KO20S2
Molecular Weight (g/mol) 499.386
MDL Number MFCD00149917
InChI Key ZFVHBEKVAITXHW-UHFFFAOYSA-J
Synonym chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
PubChem CID 24596
IUPAC Name potassium;chromium(3+);disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]

Specifications

Melting Point 89°C
Quantity 100 g
Linear Formula CrK(SO4)2·12H2O
Merck Index 14,2229
Solubility Information Soluble in water. 24g/100 mL (15°C)
Formula Weight 499.39 (283.21 Anhydrous)
Percent Purity 98%
Odor Odorless
Density 1.83
Chemical Name or Material Chromium (III) potassium sulfate dodecahydrate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 233-401-6
RTECSNumber GB6850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.