Learn More
Chromogranin A Antibody (SPM585), Alexa Fluor™ 405, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Chromogranin A |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | SPM585 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I |
| Gene Symbols | CHGA |
| Host Species | Mouse |
| Immunogen | Human pheochromocytoma (Uniprot: P10645) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.