Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromogranin C Antibody (SGII 6B1/3), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NP27661J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27661J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27661J646 Supplier Novus Biologicals Supplier No. NBP276658JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chromogranin C Monoclonal specifically detects Chromogranin C in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Chromogranin C
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone SGII 6B1/3
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CHCG, Chromogranin-C, EM66, SCG2, secretogranin IICHGCchromogranin-C, secretoneurin, SgIIsecretogranin-2, SN
Gene Symbols SCG2
Host Species Mouse
Immunogen Peptide from rat Chromogranin C (C terminus of p18: KEENSRENPF)
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7857
Target Species Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.