Learn More
CHST2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | CHST2 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5172
![]()
|
Proteintech Group Inc
260271AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody recognizes both short form and long form, which migrates around 50-52 kDa and 58-60 kDa, respectively.
N-acetylglucosamine-6-O-sulfotransferases, such as CHST2, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate.Specifications
| CHST2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q80WV3, Q9Y4C5 | |
| 54371, 9435 | |
| CHST2 Fusion Protein Ag23293 | |
| Primary | |
| -20°C | |
| CHST2 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C6ST, CHST2, GlcNAc6ST 1, GN6ST, Gn6ST 1, GST 2, GST2 | |
| Chst2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.