Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHT1 Antibody (62-2E8), Novus Biologicals™
SDP

Catalog No. NB11074570T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11074570T 0.025 mL
NB11074570 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11074570T Supplier Novus Biologicals Supplier No. NB11074570SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

CHT1 Monoclonal specifically detects CHT1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CHT1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 62-2E8
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:500, Immunohistochemistry-Paraffin 1:500, Immunohistochemistry-Frozen 1:500
Formulation 50% Glycerol with 0.05% Sodium Azide
Gene Accession No. Q9GZV3
Gene Alias CHT, CHT1MGC126299, hCHT, Hemicholinium-3-sensitive choline transporter, high affinity choline transporter 1, high affinity choline transporter; hemicholinium-3-sensitive cholinetransporter, MGC126300, solute carrier family 5 (choline transporter), member 7, Solute carrier family 5 member 7
Gene Symbols SLC5A7
Host Species Mouse
Immunogen Recombinant fusion protein from the C-terminal portion of the human CHT1 protein. [UniProt# Q9GZV3]
Purification Method Tissue culture supernatant
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 60482
Test Specificity Human, mouse and rat. Primate reactivity reported in scientific literature (PMID: 12820166).
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.