Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChT1 antigen Antibody (CT1), FITC, Novus Biologicals™
SDP

Catalog No. NBP128356 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128356 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128356 Supplier Novus Biologicals Supplier No. NBP128356
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ChT1 antigen Monoclonal specifically detects ChT1 antigen in Avian, Chicken samples. It is validated for Flow Cytometry, Immunoprecipitation, In vitro assay.

Specifications

Antigen ChT1 antigen
Applications Flow Cytometry, Immunoprecipitation, In vitro Assay
Classification Monoclonal
Clone CT1
Concentration 0.5mg/mL
Conjugate FITC
Dilution Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500, In vitro assay
Gene Alias ChT1 thymocyte antigen, V set and immunoglobulin domain containing, VSIG1
Host Species Mouse
Immunogen Chicken thymocytes
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Chicken/Quail ChT1
Target Species Avian, Chicken
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.