Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHTF18 Antibody, Novus Biologicals™
SDP

Catalog No. NB393206 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393206 25 μL
NBP258935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB393206 Supplier Novus Biologicals Supplier No. NBP25893525UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHTF18 Polyclonal specifically detects CHTF18 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CHTF18
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide
Gene Alias C16orf41, C321D2.2, C321D2.3, C321D2.3 (novel protein), C321D2.4 (novel protein), CHL12C321D2.4, chromosome 16 open reading frame 41, chromosome transmission fidelity protein 18 homolog, CTF18, CTF18, chromosome transmission fidelity factor 18 homolog (S. cerevisiae), EC 3.6.1, hCTF18, homolog of yeast CHL12, RUVBL, some homology with holliday junction DNA helicase RUVB like
Gene Symbols CHTF18
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GIGEKGVHRPAPRNHEQRLEHIMRRAAREEQPEKDFFGRVVVRSTAVPSAGDTAPEQDSVERRMGTAVGRSEVWFRFNE
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63922
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.