Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHTF18 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CHTF18 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CHTF18 Polyclonal specifically detects CHTF18 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CHTF18 | |
Polyclonal | |
Rabbit | |
Human | |
63922 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GIGEKGVHRPAPRNHEQRLEHIMRRAAREEQPEKDFFGRVVVRSTAVPSAGDTAPEQDSVERRMGTAVGRSEVWFRFNE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
C16orf41, C321D2.2, C321D2.3, C321D2.3 (novel protein), C321D2.4 (novel protein), CHL12C321D2.4, chromosome 16 open reading frame 41, chromosome transmission fidelity protein 18 homolog, CTF18, CTF18, chromosome transmission fidelity factor 18 homolog (S. cerevisiae), EC 3.6.1, hCTF18, homolog of yeast CHL12, RUVBL, some homology with holliday junction DNA helicase RUVB like | |
CHTF18 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title