Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHTF8 Antibody, Novus Biologicals™
SDP

Catalog No. NB435480 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435480 25 μL
NBP256494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435480 Supplier Novus Biologicals Supplier No. NBP25649425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHTF8 Polyclonal specifically detects CHTF8 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CHTF8
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias chromosome transmission fidelity protein 8 homolog, CTF8, chromosome transmission fidelity factor 8 homolog (S. cerevisiae), CTF8hCTF8, Decreased expression in renal and prostate cancer protein, DERPCchromosome transmission fidelity factor 8 homolog, FLJ20400
Gene Symbols CHTF8
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MGPGSGPNLRAGVLLTSGNGPPNPRPVGLGPGPNPNLRSGFLGTNPAPRSGVFPGPG
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54921
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.