Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHX10 Sheep anti-Human, Mouse, Rat, Chicken, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151736 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151736 25 μg
NB151735 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151736 Supplier Novus Biologicals Supplier No. NBP31189125UG
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

CHX10 Polyclonal antibody specifically detects CHX10 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Antigen CHX10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias ceh-10 homeo domain containing homolog, ceh-10 homeo domain containing homolog (C. elegans), ceh-10 homeodomain containing homolog (C. elegans), Ceh-10 homeodomain-containing homolog, CHX10MCOPCB3, Homeobox protein CHX10, HOX10C elegans ceh-10 homeo domain-containing homolog, MCOP2, RET1, visual system homeobox 2
Host Species Sheep
Immunogen Recombinant protein corresponding to amino acids 264 to 361 derived from the C-terminal region of the human CHX10 protein conjugated to KLH. (Uniprot: P58304)
Purification Method Ammonium sulfate precipitation
Quantity 25 μg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 338917
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.