Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chymase/CMA1/Mast Cell Chymase Rabbit anti-Human, Mouse, Rat, Clone: JB74-32, Novus Biologicals™
SDP

Catalog No. NBP275441 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275441 Supplier Novus Biologicals Supplier No. NBP275441
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Chymase/CMA1/Mast Cell Chymase Monoclonal specifically detects Chymase/CMA1/Mast Cell Chymase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Chymase/CMA1/Mast Cell Chymase
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JB74-32
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCT1, MGC119890, MGC119891
Gene Symbols CMA1
Host Species Rabbit
Immunogen Synthetic peptide within Human Chymase/CMA1/Mast Cell Chymase aa 198-247/ 247. (SwissProt: P23946 Human; SwissProt: P21844 Mouse; SwissProt: P50339 Rat)
Molecular Weight of Antigen 27 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Mast Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1215.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.