Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chymotrypsin C/CTRC Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB062975 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB062975 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB062975 Supplier Novus Biologicals Supplier No. NBP299799FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Chymotrypsin C/CTRC Polyclonal specifically detects Chymotrypsin C/CTRC in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Chymotrypsin C/CTRC
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Caldecrin, chymotrypsin C (caldecrin), chymotrypsin-C, CLCRserum calcium decreasing factor, EC 3.4.21, EC 3.4.21.2, ELA4, elastase 4, elastase IV
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Chymotrypsin C/CTRC (Q99895; Met1-Leu268).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11330
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.