Learn More
Invitrogen™ Chymotrypsin Monoclonal Antibody (CHYMO10-104.10), Invitrogen™
Description
MA1-10624 detects Chymotrypsin from human samples. MA1-10624 has been successfully used in ELISA and Western blot applications. The MA1-10624 immunogen is purified human pancreatic Chymotrypsin.
Specifications
Specifications
| Antigen | Chymotrypsin |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | CHYMO10-104.10 |
| Concentration | 2.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CTRC |
| Gene Accession No. | Q99895 |
| Gene Alias | 1810044E12Rik; caldecrin; chymotrypsin C; chymotrypsin C (caldecrin); chymotrypsin-C; CLCR; Ctrc; ELA4; elastase 4; elastase IV; preprocaldecrin; serum calcium decreasing factor; serum calcium-decreasing factor |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.