Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIAO1 Antibody (6C9), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP242631AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242631AI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242631AI Supplier Novus Biologicals Supplier No. NBP242631AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CIAO1 Monoclonal specifically detects CIAO1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CIAO1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 6C9
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CIA1, cytosolic iron-sulfur protein assembly 1, cytosolic iron-sulfur protein assembly 1 homolog (S. cerevisiae), probable cytosolic iron-sulfur protein assembly protein CIAO1, WD repeat domain 39, WD repeat-containing protein 39, WD40 protein Ciao1, WDR39cytosolic iron-sulfur protein assembly 1 homolog
Gene Symbols CIAO1
Host Species Mouse
Immunogen Recombinant human CIAO1 (1-339aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9391
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.