Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cIAP-1/HIAP-2 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB216958 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB216958 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB216958 Supplier Novus Biologicals Supplier No. NBP177193V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

cIAP-1/HIAP-2 Polyclonal antibody specifically detects cIAP-1/HIAP-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen cIAP-1/HIAP-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias API1Hiap-2, apoptosis inhibitor 1, baculoviral IAP repeat containing 2, baculoviral IAP repeat-containing 2, baculoviral IAP repeat-containing protein 2, cIAP1, C-IAP1, hIAP2, hIAP-2, IAP homolog B, IAP2, IAP-2, Inhibitor of apoptosis protein 2, MIHBHIAP2, NFR2-TRAF signalling complex protein, RING finger protein 48, RNF48hiap-2, TNFR2-TRAF-signaling complex protein 2
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human cIAP-1/HIAP-2. c-IAP antibody detects both c-IAP1 and c-IAP2. The immunogen is located within amino acids 540-590 of cIAP-1/HIAP-2. Amino Acid Sequence: SLEEQLRRLQEERT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 329
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.