Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cIAP-1/HIAP-2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056128T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056128T 0.025 mL
NB10056128 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056128T Supplier Novus Biologicals Supplier No. NB10056128SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

cIAP-1/HIAP-2 Polyclonal specifically detects cIAP-1/HIAP-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen cIAP-1/HIAP-2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Whole antisera with 0.05% Sodium Azide
Gene Alias API1Hiap-2, apoptosis inhibitor 1, baculoviral IAP repeat containing 2, baculoviral IAP repeat-containing 2, baculoviral IAP repeat-containing protein 2, cIAP1, C-IAP1, hIAP2, hIAP-2, IAP homolog B, IAP2, IAP-2, Inhibitor of apoptosis protein 2, MIHBHIAP2, NFR2-TRAF signalling complex protein, RING finger protein 48, RNF48hiap-2, TNFR2-TRAF-signaling complex protein 2
Gene Symbols BIRC2
Host Species Rabbit
Immunogen A small peptide from within amino acids 100 to 200 of human cIAP1 was used as immunogen, GenBank no. gi 2497238.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 329
Test Specificity cIAP1 has 618 amino acids and migrates at ∼70 kDa on SDS-PAGE. Observed molecular weights may vary as genes generally produce, by alternative splicing, multiple transcripts all with introns, putatively encoding multiple different protein isoforms. Additionally, proteins may undergo post translational modifications including phosphorylation, glycosylation and cleavage which can result in differential molecular weights. Protein modifications can vary between cell and tissue type, and proteins can migrate as one or multiple bands depending on the isoform expressed as well as post-translational modifications.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.