Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cIAP-1/HIAP-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $316.50
Specifications
Antigen | cIAP-1/HIAP-2 |
---|---|
Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056128T
![]() |
Novus Biologicals
NB10056128SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB10056128
![]() |
Novus Biologicals
NB10056128 |
0.05 mL |
Each for $316.50
|
|
|||||
Description
cIAP-1/HIAP-2 Polyclonal specifically detects cIAP-1/HIAP-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
cIAP-1/HIAP-2 | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis | |
Whole antisera with 0.05% Sodium Azide | |
329 | |
A small peptide from within amino acids 100 to 200 of human cIAP1 was used as immunogen, GenBank no. gi 2497238. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
API1Hiap-2, apoptosis inhibitor 1, baculoviral IAP repeat containing 2, baculoviral IAP repeat-containing 2, baculoviral IAP repeat-containing protein 2, cIAP1, C-IAP1, hIAP2, hIAP-2, IAP homolog B, IAP2, IAP-2, Inhibitor of apoptosis protein 2, MIHBHIAP2, NFR2-TRAF signalling complex protein, RING finger protein 48, RNF48hiap-2, TNFR2-TRAF-signaling complex protein 2 | |
BIRC2 | |
IgG | |
Unpurified | |
cIAP1 has 618 amino acids and migrates at ∼70 kDa on SDS-PAGE. Observed molecular weights may vary as genes generally produce, by alternative splicing, multiple transcripts all with introns, putatively encoding multiple different protein isoforms. Additionally, proteins may undergo post translational modifications including phosphorylation, glycosylation and cleavage which can result in differential molecular weights. Protein modifications can vary between cell and tissue type, and proteins can migrate as one or multiple bands depending on the isoform expressed as well as post-translational modifications. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title