Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cIAP-2/HIAP-1 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP127972R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP127972R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP127972R Supplier Novus Biologicals Supplier No. NBP127972R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

cIAP-2/HIAP-1 Polyclonal specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen cIAP-2/HIAP-1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q13489
Gene Alias AIP1, API2, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49
Gene Symbols BIRC3
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human cIAP2 (within residues 100-200). [Swiss-Prot# Q13489]
Molecular Weight of Antigen 64 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 330
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.