Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cIAP-2/HIAP-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056132 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056132 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056132 Supplier Novus Biologicals Supplier No. NB10056132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

cIAP-2/HIAP-1 Polyclonal specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for IF/IHC, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen cIAP-2/HIAP-1
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen 1:10-1:500
Formulation Neat whole antisera with 0.05% Sodium Azide
Gene Accession No. Q13489
Gene Alias AIP1, API2, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49
Gene Symbols BIRC3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 152-172 (QDFSALMRSSYHCAMNNENAR) of human cIAP2 was used as immunogen (NP_892007).
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 330
Test Specificity cIAP2
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.