Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cIAP-2/HIAP-1 Antibody (OTI1A9), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP270184A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270184A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270184A94 Supplier Novus Biologicals Supplier No. NBP270184AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

cIAP-2/HIAP-1 Monoclonal specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen cIAP-2/HIAP-1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1A9
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AIP1, API2, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49
Gene Symbols BIRC3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 255-529 of human cIAP-2/HIAP-1 (NP_001156) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 330
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.