Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cIAP-2/HIAP-1 Antibody (OTI1A9), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N270184J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270184J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270184J549 Supplier Novus Biologicals Supplier No. NBP270184JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

cIAP-2/HIAP-1 Monoclonal specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen cIAP-2/HIAP-1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1A9
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AIP1, API2, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49
Gene Symbols BIRC3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 255-529 of human cIAP-2/HIAP-1 (NP_001156) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Inhibitors Activators and Regulators, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 330.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.