Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cIAP1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1H3F1, Proteintech
SDP

Catalog No. 501737044 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7044 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7044 Supplier Proteintech Group Inc Supplier No. 666261IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

cIAP1 encodes a protein that is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. This encoded protein inhibits apoptosis induced by serum deprivation and menadione, a potent inducer of free radicals.

Specifications

Antigen cIAP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1H3F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene BIRC2
Gene Accession No. Q13490, Q62210
Gene Alias API1, BIRC2, C IAP1, cIAP1, Hiap 2, HIAP2, IAP 2, IAP homolog B, IAP2, MIHB, RING finger protein 48, RNF48
Gene Symbols BIRC2
Host Species Mouse
Immunogen cIAP1 Fusion Protein Ag21398
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100622859, 11797, 329, 60371
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.