Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIAPIN1 Antibody (OTI5A6), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP272072A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272072A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272072A32 Supplier Novus Biologicals Supplier No. NBP272072AF532

Mouse Monoclonal Antibody

CIAPIN1 Monoclonal specifically detects CIAPIN1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CIAPIN1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5A6
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Anamorsin, cytokine induced apoptosis inhibitor 1, Cytokine-induced apoptosis inhibitor 1, DRE2, Fe-S cluster assembly protein DRE2 homolog, predicted protein of HQ0915,2810413N20Rik, PRO0915
Gene Symbols CIAPIN1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 97-312 of human CIAPIN1 (NP_064709) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57019
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.