Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIB1 Antibody (1D1), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP104279UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104279UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104279UV Supplier Novus Biologicals Supplier No. NBP104279UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CIB1 Monoclonal specifically detects CIB1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CIB1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1D1
Conjugate DyLight 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias calcium and integrin binding 1 (calmyrin), Calcium- and integrin-binding protein, calmyrin, CIBcalcium and integrin binding protein, CIBP, DNA-dependent protein kinase interacting protein, DNA-PKcs-interacting protein, Kinase-interacting protein, KIP1, KIPcalcium and integrin binding, protein kinase interacting protein, PRKDCIP, SIP2-28calcium and integrin-binding protein 1, Snk interacting protein 2-28, SNK-interacting protein 2-28
Gene Symbols CIB1
Host Species Mouse
Immunogen Recombinant human CIB1 (1-191aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10519
Test Specificity Anti-human CIB1 mAb, clone 1D1, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CIB1 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.