Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIB3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB170035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170035 100 μg
NB170036 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB170035 Supplier Novus Biologicals Supplier No. NBP317480100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CIB3 Polyclonal antibody specifically detects CIB3 in Human samples. It is validated for Immunofluorescence

Specifications

Antigen CIB3
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias calcium and integrin binding family member 3, calcium and integrin-binding family member 3, DNA-dependent protein kinase catalytic subunit-interacting protein 3, Kinase-interacting protein 3, KIP 3, KIP3MGC96922, MGC138405, MGC142151
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: LEAYQDCTFFTRKEIMRLFYRYQDLAPQLVPLDYTTCPDVKVPYELIGSMPELKDNPFRQRIAQVFSEDGDGHMTLDNFLDMFSVMSEMAPR
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117286
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.