Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cig2 Antibody (3A11/5), HRP, Novus Biologicals™
SDP

Catalog No. NB392289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392289 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392289 Supplier Novus Biologicals Supplier No. NBP308443H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cig2 Monoclonal antibody specifically detects Cig2 in S. pombe samples. It is validated for Western Blot, ELISA

Specifications

Antigen Cig2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3A11/5
Conjugate HRP
Formulation PBS
Host Species Mouse
Immunogen Bacterial S. pombe Cig2 Purified protein.
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Target Species S. pombe
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.