Learn More
Ciglitazone, Tocris
Selective PPARγ agonist
Manufacturer:  R&D Systems 1307/10
Quantity 10mg
 View more versions of this product
Catalog No. 13-071-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
333.44
Ciglitazone
C18H23NO3S
5-[[4-[(1-Methylcyclohexyl)methoxy]phenyl]methyl]-2, 4-thiazolidinedione
Specifications
74772-77-3
C18H23NO3S
5-[[4-[(1-Methylcyclohexyl)methoxy]phenyl]methyl]-2, 4-thiazolidinedione
Ciglitazone
>99%
333.44
10mg
Safety and Handling
Recommended Storage : Store at Room Temperature