Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIITA Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB202578 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB202578 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB202578 Supplier Novus Biologicals Supplier No. NBP259072JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CIITA Polyclonal antibody specifically detects CIITA in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CIITA
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias C2TA, C2TAMHC class II transactivator type III, CIITAIV, class II, major histocompatibility complex, transactivator, MHC class II transactivator, MHC2TANLR family, acid domain containing, NLRA, nucleotide-binding oligomerization domain, leucine rich repeat and acid domaincontaining
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human CIITA. The immunogen is located within amino acids 120 - 170 of CIITA. Amino Acid Squence: AEVCQSQRRPFPEEL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4261
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.