Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ciliobrevin A

Catalog No. 452925
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Hedgehog (Hh) pathway antagonist, inhibits ciliogenesis; also inhibits dynein

Ciliobrevin A is a Hedgehog (Hh) pathway antagonist; blocks Sonic hedgehog (Shh)-induced Hh pathway activation (IC50 = 7 μM) downstream of Smo. Perturbs primary cilia formation; inhibits cytoplasmic AAA+ ATPase dynein-dependent microtubule gliding and ATPase activity.

Specifications

Chemical Name or Material 2,4-Dichloro-α-(3,4-4-oxo-2(1H)-quinazolinylidene)-β-oxobenzenepropanenitrile
CAS 302803-72-1
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C17H9Cl2N3O2
Formula Weight Observed MW: 358.18
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.