Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CIM 0216
GREENER_CHOICE

Catalog No. p-7110840
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective TRPM3 agonist

CIM 0216 is a selective TRPM3 agonist. Exhibits selectivity for TRPM3 over TRPM1, TRPM2 and TRPM4-8. Stimulates insulin release from pancreatic islet cells in vitro. Evokes nocifensive behavior in mice.

Specifications

Chemical Name or Material 3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-α-phenyl-1(2H)-quinolineacetamide
CAS 1031496-06-6
Quantity 50 mg
Target TRPM Agonists
Molecular Formula C21H21N3O2
Formula Weight 347.41
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.