Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cimaterol

Catalog No. 043510r
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

β agonist

Cimaterol is a β-Adrenergic agonist.

Specifications

Chemical Name or Material 2-Amino-5-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]benzonitrile
CAS 54239-37-1
Assay Percent Range >99%
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C12H17N3O
Formula Weight Observed MW: 219.29

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.