Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CINAP Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB335453 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB335453 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB335453 Supplier Novus Biologicals Supplier No. NBP298094PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CINAP Polyclonal antibody specifically detects CINAP in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CINAP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CASK-interacting nucleosome assembly protein, CDA1CTCL-associated antigen se20-4, Cell division autoantigen 1, CINAPSE20-4, CTCL, CTCL tumor antigen se20-4, Cutaneous T-cell lymphoma-associated antigen se20-4, cutaneous T-cell lymphoma-associated tumor antigen se20-4, DENTTTSPY-like protein 2, differentially expressed nucleolar TGF-beta1 target, Differentially-expressed nucleolar TGF-beta1 target protein, HRIHFB2216, NP79, Nuclear protein of 79 kDa, SE204, testis-specific protein Y encoded-like 2, testis-specific Y-encoded-like protein 2, TSPX, TSPY-like 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CINAP fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 64061
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.