Learn More
Cincinnati Surgical Sterile-Stainless Steel Blades and Scalpels
Packaged with VCI Paper to protect the blade from corrosion.
$52.30 - $148.00
Specifications
Blade Material | British Stainless Steel |
---|---|
Description | Medium Surface Finish |
Sterility | Sterile |
Catalog Number | Mfr. No. | Size | Quantity | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Quantity | Product Type | Price | Quantity & Availability | |||
19-200-215
![]() |
Cincinnati Surgical
0110 |
# 10 | 10/Pk. | Blade |
Pack of 100 for $123.80
|
|
|||
19-200-216
![]() |
Cincinnati Surgical
0111 |
# 11 | 10/Pk. | Blade |
Pack of 100 for $123.80
|
|
|||
19-200-217
![]() |
Cincinnati Surgical
0112 |
# 12 | 10/Pk. | Blade |
Pack of 100 for $123.80
|
|
|||
19-200-218
![]() |
Cincinnati Surgical
0115 |
10/Pk. | Blade |
Pack of 100 for $123.80
|
|
||||
19-200-219
![]() |
Cincinnati Surgical
0120 |
# 20 | 10/Pk. | Blade |
Pack of 100 for $127.00
|
|
|||
19-200-220
![]() |
Cincinnati Surgical
0121 |
# 21 | 10/Pk. | Blade |
Pack of 100 for $127.00
|
|
|||
19-200-221
![]() |
Cincinnati Surgical
0122 |
# 22 | 10/Pk. | Blade |
Pack of 100 for $127.00
|
|
|||
19-200-204
![]() |
Cincinnati Surgical
0510 |
# 10 | 10/Pk. | Safety Scalpel |
Pack of 10 for $54.20
|
|
|||
19-200-205
![]() |
Cincinnati Surgical
0511 |
10/Pk. | Safety Scalpel |
Pack of 10 for $54.20
|
|
||||
19-200-206
![]() |
Cincinnati Surgical
0515 |
# 15 | 10/Pk. | Safety Scalpel |
Pack of 10 for $52.30
|
|
|||
19-200-208
![]() |
Cincinnati Surgical
0521 |
# 21 | 10/Pk. | Safety Scalpel |
Pack of 10 for $59.20
|
|
|||
19-200-209
![]() |
Cincinnati Surgical
0522 |
# 22 | 10/Pk. | Safety Scalpel |
Pack of 10 for $60.30
|
|
|||
19-200-252
![]() |
Cincinnati Surgical
0115C |
# 15C | 10/Pk. | Blade |
Pack of 100 for $129.30
|
|
|||
19-200-258
![]() |
Cincinnati Surgical
05S10 |
# 10 | 25/Pk. | Safety Scalpel |
Pack of 25 for $148.00
|
|
|||
Description
- Made from British stainless steel
- 200% visual inspection
- Narrow grinding angle resulting in increased initial keenness
- Medium surface finish
- Strong foil package
Specifications
British Stainless Steel | |
Medium Surface Finish | |
Sterile |