Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CINP Antibody (1G10), PE, Novus Biologicals™
SDP

Catalog No. NB308851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308851 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308851 Supplier Novus Biologicals Supplier No. NBP259486PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CINP Monoclonal antibody specifically detects CINP in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen CINP
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1G10
Conjugate PE
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias CDK2-interacting protein, cyclin-dependent kinase 2 interacting protein, cyclin-dependent kinase 2-interacting protein, MGC849
Host Species Mouse
Immunogen Recombinant human CINP (1-212aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 51550
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.