Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIP29 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB102969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102969 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102969 Supplier Novus Biologicals Supplier No. NBP297653H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CIP29 Polyclonal specifically detects CIP29 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CIP29
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias CIP29Hcc-1, cytokine induced protein 29 kDa, Cytokine-induced protein of 29 kDa, HCC1MGC14726, hepatocellular carcinoma 1, HSPC316, Nuclear protein Hcc-1, proliferation associated cytokine-inducible protein CIP29, Proliferation-associated cytokine-inducible protein CIP29, SAP domain containing ribonucleoprotein, SAP domain-containing ribonucleoprotein, THO1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CIP29 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84324
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.