Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIP2A Antibody (2G10), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N1059722AP7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1059722AP7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1059722AP7 Supplier Novus Biologicals Supplier No. NB11059722APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CIP2A Monoclonal specifically detects CIP2A in Human, Mouse, Rat, Primate samples. It is validated for Flow Cytometry, Knockdown Validated.

Specifications

Antigen CIP2A
Applications Flow Cytometry, KnockDown
Classification Monoclonal
Clone 2G10
Conjugate APC-Cyanine7
Dilution Flow Cytometry, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cancerous inhibitor of PP2A, CIP2A, FLJ12850, KIAA1524, MGC163436, p90, p90 autoantigen, protein CIP2A
Gene Symbols CIP2A
Host Species Mouse
Immunogen A synthetic peptide made to a C-terminal portion of the human CIP2A protein. [UniProt No. Q8TCG1]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 57650.0
Test Specificity p90 autoantigen (2G10)
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.