Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIP2A Antibody, Novus Biologicals™
SDP

Catalog No. NB10068264 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068264 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068264 Supplier Novus Biologicals Supplier No. NB10068264
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 11 publications

CIP2A Polyclonal specifically detects CIP2A in Human, Mouse, Rat samples. It is validated for IF/IHC, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen CIP2A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:200
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q8TCG1
Gene Alias Cancerous inhibitor of PP2A, CIP2A, FLJ12850, KIAA1524, MGC163436, p90, p90 autoantigen, protein CIP2A
Gene Symbols CIP2A
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 853 and 903 of human cancerous inhibitor of PP2A using the numbering given in entry NP_065941.1 (GeneID 57650).
Molecular Weight of Antigen 102 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57650
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant, Small-eared galago, Little brown bat and Northern white-cheeked gibbon.
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.