Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cirazoline hydrochloride
GREENER_CHOICESDP

Catalog No. 088810 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-881-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 08-881-0 Supplier Tocris Bioscience™ Supplier No. 0888/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective α1 agonist

Selective α1 agonist.

Chemical Identifiers

CAS 40600-13-3
Molecular Formula C13H17ClN2O

Specifications

Quantity 10 mg
Formula Weight 252.74
Purity >99%
Molecular Weight (g/mol) 252.742
Chemical Name or Material Cirazoline hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.