Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Circlegrow™ Agar
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
908 g
Description
- CIRCLEGROW™ Agar media is used for growth of E.coli.
- It also extends plasmid production past log phase.
- Acceptable pH range is 6.9 ± 0.3 at 25°C.
- Please note that the pH of CIRCLEGROW™ Medium liquid increases as the temperature of the solution decreases.

Specifications
Specifications
Concentration | 55 g/L |
Form | Powder |
Grade | Molecular Biology Certified™ |
Media Type | Circlegrow |
Preparation Method | Reagents are tested and chosen for their ability to promote optimum growth of E. coli when combined in Circlegrow™ Agar formulations. Chosen reagents are combined in the accurate proportions according to the proprietary Circlegrow™ Agar formula and thoroughly blended for a uniform distribution. |
Formulation | Proprietary formulation containing Yeast Extract, Tryptone Broth, and Vitamins. Biological buffers have been added for proper pH adjustment. |
For Use With (Application) | Cell Culture, Proteomics, Purification and Detection, Recombinant Protein Expression and Analysis, Bacterial Culture, Plasmid Production, Protein Expression, Gene Expression, Cloning, Genetic Manipulation |
Quantity | 908 g |
Solubility | Soluble in water, 55 g/L |
Synonym | Enhanced Bacterial Media |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction