Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cis-Vaccenic Acid Analytical Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
100 mg
Specifications
Specifications
| CAS | 506-17-2 |
| Melting Point | 14°C to 15°C (literature) |
| Density | 0.887 g/mL (at 25°C (literature)) |
| Boiling Point | 150°C/0.03 mmHg (literature) |
| Flash Point | 230°C (Closed Cup) |
| Molecular Formula | C18H34O2 |
| Linear Formula | CH3(CH2)5 CH=CH(CH2)9 CO2H |
| Refractive Index | n20/D 1.459 (literature) |
| Quantity | 100 mg |
| MDL Number | MFCD00063187 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction