Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Zeatin, MP Biomedicals™

Catalog No. p-4679580 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg

Chemical Identifiers

CAS 32771-64-5
Molecular Formula C10H13N5O
Molecular Weight (g/mol) 219.248
MDL Number MFCD00070217
InChI Key UZKQTCBAMSWPJD-UQCOIBPSSA-N
Synonym cis-zeatin, 2z-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, z-2-methyl-4-1h-purin-6-ylamino but-2-en-1-ol, z-4-7h-purin-6-yl amino-2-methylbut-2-en-1-ol, 6-4-hydroxy-3-methyl-cis-2-butenylamino purine, e-zeatin, 2z-2-methyl-4-9h-purin-6-ylamino-2-buten-1-ol, cis-zeatin hplc, z-2-methyl-4-7h-purin-6-ylamino but-2-en-1-ol
PubChem CID 688597
ChEBI CHEBI:46570
IUPAC Name (Z)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol
SMILES CC(=CCNC1=NC=NC2=C1NC=N2)CO

Specifications

Quantity 1 mg
Formula Weight 219.2
Physical Form Solid
Chemical Name or Material cis-Zeatin
Recommended Storage Store at -0°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.