Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CISD1 Antibody (OTI4G3), PE, Novus Biologicals™
SDP

Catalog No. NB288427 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288427 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288427 Supplier Novus Biologicals Supplier No. NBP272026PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CISD1 Monoclonal antibody specifically detects CISD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen CISD1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone OTI4G3
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias C10orf70, CDGSH iron sulfur domain 1, CDGSH iron sulfur domain-containing protein 1, CDGSH iron-sulfur domain-containing protein 1, chromosome 10 open reading frame 70, MDS029, MGC14684, mitoNEET, ZCD1, zinc finger CDGSH-type domain 1, zinc finger, CDGSH-type domain 1
Host Species Mouse
Immunogen Full length human recombinant protein of human CISD1 (NP_060934) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 55847
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.