Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cisplatin Modified DNA Antibody (CP9/19), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP250165AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250165AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250165AI Supplier Novus Biologicals Supplier No. NBP250165AF700
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Cisplatin Modified DNA Monoclonal specifically detects Cisplatin Modified DNA in Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cisplatin Modified DNA
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CP9/19
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ICR4
Host Species Rat
Immunogen Cisplatin modified native DNA
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics
Primary or Secondary Primary
Test Specificity CP9/19 recognises only the intra-strand cross-links formed by Cisplatin between adjacent purines.
Target Species Other
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.