Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Citalopram hydrobromide
GREENER_CHOICE

Catalog No. p-4783356
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly potent and selective 5-HT uptake inhibitor

Citalopram hydrobromide is a highly selective and potent 5-HT uptake inhibitor with no effect on noradrenalin or dopamine uptake (IC50 values are 1.8, 8800 and 41000 nM respectively). Has negligible activity at a wide range of receptors.

Chemical Identifiers

CAS 59729-32-7
Molecular Formula C20H22BrFN2O

Specifications

Quantity 10 mg
Formula Weight 405.31
Purity >99%
Molecular Weight (g/mol) 405.311
Chemical Name or Material Citalopram hydrobromide
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.