Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cited-2 Antibody (JA22), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB001686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001686 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001686 Supplier Novus Biologicals Supplier No. NB100136IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cited-2 Monoclonal specifically detects Cited-2 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Cited-2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone JA22
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain2,cbp/p300-interacting transactivator 2, MRG-1, MRG1melanocyte-specific gene 1-related gene 1, MSG1-related gene 1, MSG-related protein 1, P35srj
Gene Symbols CITED2
Host Species Mouse
Immunogen Human CITED 2 protein [UniProt# Q99967]
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Chromatin Research, HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 10370
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.