Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CITED4 Antibody (HT13-2D6.3), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1104172A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1104172A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1104172A32 Supplier Novus Biologicals Supplier No. NB11041572AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CITED4 Monoclonal specifically detects CITED4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CITED4
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HT13-2D6.3
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cbp/p300-interacting transactivator 4, Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain4,MRG-2, MRG2, MSG1-related protein 2, transcriptional co-activator 4
Gene Symbols CITED4
Host Species Mouse
Immunogen N-terminal fusion protein to full-length human CITED4 [UniProtNo. Q96RK1]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 163732
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.