Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CITED4 Antibody (HT13-2D6.3), PE, Novus Biologicals™
SDP

Catalog No. NB11041572Q Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11041572Q 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11041572Q Supplier Novus Biologicals Supplier No. NB11041572PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CITED4 Monoclonal specifically detects CITED4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CITED4
Applications Flow Cytometry
Classification Monoclonal
Clone HT13-2D6.3
Conjugate PE
Dilution Flow Cytometry
Gene Accession No. Q96RK1
Gene Alias cbp/p300-interacting transactivator 4, Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain4,MRG-2, MRG2, MSG1-related protein 2, transcriptional co-activator 4
Gene Symbols CITED4
Host Species Mouse
Immunogen N-terminal fusion protein to full-length human CITED4 [UniProt# Q96RK1]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 163732
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.