Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citrate, 0.5 M Buffer Solution, pH 5.0, PROMO, Thermo Scientific Chemicals

Catalog No. J60125APPM
Click to view available options
Quantity:
500 mL

Applications

Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.

Solubility

It is soluble in water (29.4 g/L) at 20°C. Insoluble in ethanol.

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6132-04-3

Specifications

Color Colorless
Concentration 0.5 M
Molecular Weight (g/mol) 294.1 g/mol
pH 5.0
Quantity 500 mL
Content And Storage Keep cold
Form Liquid

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.