Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, 1.00 M, Ricca Chemical
SDP

Catalog No. p-3251504 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2131-16 500 mL Natural Poly Bottle
2131-32 1 L Natural Poly Bottle
21311 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2131-16 Supplier Ricca Chemical Company Supplier No. 213116
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

CAS Min % 82.0, 15.8
Color Colorless
CAS Max % 86.0, 16.45
Concentration 1.00 Molar
Grade Laboratory
Quantity 500 mL
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Citric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.