Learn More
Citric acid, Electrophoresis Grade, 99.5+%
Description
Citric acid is a starting point in the Krebs cycle and is a key intermediate in metabolism. It has anticoagulant activity; as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot. It may be used to adjust pH and as a sequestering agent for the removal of trace metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Citric acid is a starting point in the Krebs cycle and is a key intermediate in metabolism. It has anticoagulant activity; as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot. It may be used to adjust pH and as a sequestering agent for the removal of trace metals.
Solubility
Soluble in water. Also soluble in alcohol and methanol. Insoluble in chloroform and benzene.
Notes
Hygroscopic. Incompatible with oxidizing agents.

Specifications
Specifications
pH | 1.6 |
Melting Point | 153°C to 154°C |
Color | White |
Beilstein | 782061 |
Merck Index | 14,2326 |
Solubility Information | Soluble in water. Also soluble in alcohol and methanol. Insoluble in chloroform and benzene. |
Formula Weight | 192.13 |
Percent Purity | ≥99.5% |
Grade | Electrophoresis |
Density | 1.665 |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | H319-H335-H500 |
missing translation for 'healthHazard2' | GHS H Statement H318-H335-H315 Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
missing translation for 'healthHazard3' | P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c |
EINECSNumber | 201-069-1 |
RTECSNumber | GE7350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.